Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAATATGTTGAAGCATTAATTA[A/C]CTTGAAACAAAAAATTATCAATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ICE1 - interactor of little elongation complex ELL subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015325.2 | 359 | Missense Mutation | AAC,ACC | N,T 39 | NP_056140.1 | |
XM_011513999.2 | 359 | Missense Mutation | AAC,ACC | N,T 39 | XP_011512301.1 | |
XM_017009285.1 | 359 | UTR 5 | XP_016864774.1 |