Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCAGATACCCCCTCCTCACCCT[C/G]GTCTTCCAATGAGGAAGTAGTATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615769 MIM: 612497 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM169A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM169A - family with sequence similarity 169 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015566.2 | 2024 | Missense Mutation | CAG,GAG | Q,E 571 | NP_056381.1 | |
XM_005248480.2 | 2024 | Missense Mutation | CAG,GAG | Q,E 571 | XP_005248537.1 | |
XM_011543306.2 | 2024 | Missense Mutation | CAG,GAG | Q,E 571 | XP_011541608.1 | |
XM_011543307.2 | 2024 | Missense Mutation | CAG,GAG | Q,E 571 | XP_011541609.1 | |
XM_017009332.1 | 2024 | Missense Mutation | CAG,GAG | Q,E 547 | XP_016864821.1 | |
XM_017009333.1 | 2024 | Missense Mutation | CAG,GAG | Q,E 485 | XP_016864822.1 |
NSA2 - NSA2, ribosome biogenesis homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |