Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAACTCTCTTCTTCGCCAAACAT[A/G]TCTGCCAAGACTTTAAAGCGGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH12 - cadherin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317227.1 | 3054 | Silent Mutation | GAC,GAT | D,D 780 | NP_001304156.1 | |
NM_001317228.1 | 3054 | Silent Mutation | GAC,GAT | D,D 740 | NP_001304157.1 | |
NM_004061.4 | 3054 | Silent Mutation | GAC,GAT | D,D 780 | NP_004052.2 | |
XM_011513927.2 | 3054 | Silent Mutation | GAC,GAT | D,D 780 | XP_011512229.1 | |
XM_017008920.1 | 3054 | Silent Mutation | GAC,GAT | D,D 780 | XP_016864409.1 | |
XM_017008921.1 | 3054 | Silent Mutation | GAC,GAT | D,D 780 | XP_016864410.1 |