Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTTCTGGAGAAAGTGGCTATGCT[C/T]AAAATGGTGAGTCATTTCTAAGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146661 | ||||||||||||||||||||
Literature Links: |
IL7R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL7R - interleukin 7 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002185.3 | 164 | Nonsense Mutation | CAA,TAA | Q,* 26 | NP_002176.2 | |
XM_005248299.3 | 164 | Nonsense Mutation | CAA,TAA | Q,* 26 | XP_005248356.1 |