Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCAGCGGCCGCGGGCGGCGGCC[C/G]GCCCGCAGGGAAGTCCGGGGCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602406 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAND1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAND1 - heart and neural crest derivatives expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004821.2 | 379 | Missense Mutation | CGG,GGG | R,G 60 | NP_004812.1 | |
XM_005268531.1 | 379 | Missense Mutation | CGG,GGG | R,G 60 | XP_005268588.1 |