Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGGGCCCCCGCATCATTGGCT[C/T]AAGGCATGAAATCTCAGTTGAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAPT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAPT - GRB2-binding adaptor protein, transmembrane | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304428.1 | 731 | Missense Mutation | TCA,TTA | S,L 69 | NP_001291357.1 | |
NM_001304429.1 | 731 | Missense Mutation | TCA,TTA | S,L 69 | NP_001291358.1 | |
NM_001304431.1 | 731 | Missense Mutation | TCA,TTA | S,L 69 | NP_001291360.1 | |
NM_152687.3 | 731 | Missense Mutation | TCA,TTA | S,L 69 | NP_689900.1 |