Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATGGCTGTATCTACCGCAGACT[A/G]AGGAATCATCCCACGCAGATCTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114080 MIM: 607049 | ||||||||||||||||||||
Literature Links: |
CAMK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAMK4 - calcium/calmodulin dependent protein kinase IV | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STARD4 - StAR related lipid transfer domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308056.1 | 769 | Nonsense Mutation | CAG,TAG | Q,* 183 | NP_001294985.1 | |
NM_001308057.1 | 769 | Intron | NP_001294986.1 | |||
NM_001308058.1 | 769 | Intron | NP_001294987.1 | |||
NM_001308059.1 | 769 | Nonsense Mutation | CAG,TAG | Q,* 101 | NP_001294988.1 | |
NM_001308060.1 | 769 | Nonsense Mutation | CAG,TAG | Q,* 85 | NP_001294989.1 | |
NM_001308061.1 | 769 | Nonsense Mutation | CAG,TAG | Q,* 101 | NP_001294990.1 | |
NM_139164.2 | 769 | Nonsense Mutation | CAG,TAG | Q,* 183 | NP_631903.1 | |
XM_017009041.1 | 769 | Nonsense Mutation | CAG,TAG | Q,* 85 | XP_016864530.1 | |
XM_017009042.1 | 769 | Nonsense Mutation | CAG,TAG | Q,* 85 | XP_016864531.1 | |
XM_017009043.1 | 769 | Nonsense Mutation | CAG,TAG | Q,* 85 | XP_016864532.1 | |
XM_017009044.1 | 769 | Nonsense Mutation | CAG,TAG | Q,* 101 | XP_016864533.1 |