Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCAGCATCGGCTTCAGCAAGAA[A/C]GACTTCGTGATGCGTGCCCGCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600366 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISL1 - ISL LIM homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002202.2 | 815 | Missense Mutation | AAA,AAC | K,N 89 | NP_002193.2 | |
XM_011543380.2 | 815 | Missense Mutation | AAA,AAC | K,N 25 | XP_011541682.1 |
LOC642366 - uncharacterized LOC642366 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |