Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTTTGGAAGAATTGAATACCTC[G/T]TTGGTGTTGAGCTTGATGTCTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf51 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf51 - chromosome 5 open reading frame 51 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO4 - F-box protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297437.1 | 637 | Silent Mutation | TCG,TCT | S,S 192 | NP_001284366.1 | |
NM_012176.2 | 637 | Silent Mutation | TCG,TCT | S,S 192 | NP_036308.1 | |
NM_033484.2 | 637 | Silent Mutation | TCG,TCT | S,S 192 | NP_277019.1 | |
XM_006714470.3 | 637 | Silent Mutation | TCG,TCT | S,S 48 | XP_006714533.1 | |
XM_011514026.2 | 637 | Silent Mutation | TCG,TCT | S,S 192 | XP_011512328.1 | |
XM_011514027.2 | 637 | Silent Mutation | TCG,TCT | S,S 58 | XP_011512329.1 |