Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCGACTTGAAGGACTCCAGGCG[A/C]TTCCGCATGGTCCTGTGGAAGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608186 MIM: 182307 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOC3 - exocyst complex component 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100288152 - uncharacterized LOC100288152 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PP7080 - uncharacterized LOC25845 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9A3 - solute carrier family 9 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284351.2 | 2089 | Missense Mutation | AAG,AAT | K,N 650 | NP_001271280.1 | |
NM_004174.3 | 2089 | Missense Mutation | AAG,AAT | K,N 659 | NP_004165.2 |