Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTTCAGTTTCTTCTTTTTCTCT[C/T]TGTTGGAGAGTTTGGCATCTGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEPDC1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEPDC1B - DEP domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145208.1 | 1469 | Intron | NP_001138680.1 | |||
NM_018369.2 | 1469 | Missense Mutation | AAA,AGA | K,R 468 | NP_060839.2 | |
XM_011543509.2 | 1469 | Missense Mutation | AAA,AGA | K,R 453 | XP_011541811.1 | |
XM_011543510.2 | 1469 | Intron | XP_011541812.1 | |||
XM_017009629.1 | 1469 | Intron | XP_016865118.1 |