Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGAGGGACTTTCCCTTTCGTCCG[C/G]CGAAGGTCTGGTGAGTCCAATGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEF2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEF2C - myocyte enhancer factor 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131005.2 | 1475 | Missense Mutation | CCG,GCG | P,A 444 | NP_001124477.1 | |
NM_001193347.1 | 1475 | Missense Mutation | CCG,GCG | P,A 464 | NP_001180276.1 | |
NM_001193348.1 | 1475 | Missense Mutation | CCG,GCG | P,A 398 | NP_001180277.1 | |
NM_001193349.1 | 1475 | Missense Mutation | CCG,GCG | P,A 374 | NP_001180278.1 | |
NM_001193350.1 | 1475 | Missense Mutation | CCG,GCG | P,A 454 | NP_001180279.1 | |
NM_001308002.1 | 1475 | Missense Mutation | CCG,GCG | P,A 446 | NP_001294931.1 | |
NM_002397.4 | 1475 | Missense Mutation | CCG,GCG | P,A 454 | NP_002388.2 | |
XM_005248511.2 | 1475 | Missense Mutation | CCG,GCG | P,A 454 | XP_005248568.1 | |
XM_006714619.2 | 1475 | Missense Mutation | CCG,GCG | P,A 454 | XP_006714682.1 | |
XM_006714625.3 | 1475 | Missense Mutation | CCG,GCG | P,A 464 | XP_006714688.1 | |
XM_011543396.2 | 1475 | Missense Mutation | CCG,GCG | P,A 454 | XP_011541698.1 | |
XM_011543397.2 | 1475 | Missense Mutation | CCG,GCG | P,A 452 | XP_011541699.1 | |
XM_011543400.1 | 1475 | Missense Mutation | CCG,GCG | P,A 422 | XP_011541702.1 | |
XM_011543401.1 | 1475 | Missense Mutation | CCG,GCG | P,A 414 | XP_011541703.1 | |
XM_017009475.1 | 1475 | Missense Mutation | CCG,GCG | P,A 452 | XP_016864964.1 | |
XM_017009476.1 | 1475 | Missense Mutation | CCG,GCG | P,A 446 | XP_016864965.1 | |
XM_017009477.1 | 1475 | Missense Mutation | CCG,GCG | P,A 446 | XP_016864966.1 | |
XM_017009478.1 | 1475 | Missense Mutation | CCG,GCG | P,A 444 | XP_016864967.1 | |
XM_017009479.1 | 1475 | Missense Mutation | CCG,GCG | P,A 422 | XP_016864968.1 | |
XM_017009480.1 | 1475 | Missense Mutation | CCG,GCG | P,A 422 | XP_016864969.1 | |
XM_017009481.1 | 1475 | Missense Mutation | CCG,GCG | P,A 422 | XP_016864970.1 | |
XM_017009482.1 | 1475 | Missense Mutation | CCG,GCG | P,A 414 | XP_016864971.1 | |
XM_017009483.1 | 1475 | Missense Mutation | CCG,GCG | P,A 412 | XP_016864972.1 |