Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGGCCCTCCCCGTGCCTGCCCCC[C/T]AGAGCCGCCCCCTGAGCACTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALT7 - beta-1,4-galactosyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007255.2 | 2446 | Missense Mutation | CCA,CTA | P,L 77 | NP_009186.1 | |
XM_006714816.3 | 2446 | UTR 5 | XP_006714879.1 | |||
XM_017008999.1 | 2446 | UTR 5 | XP_016864488.1 |
TMED9 - transmembrane p24 trafficking protein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |