Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGTATGCCCAACCCCAGTTCTC[C/T]GTGTTCCTCGGTCTCATCTTCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EIF4E1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF4E1B - eukaryotic translation initiation factor 4E family member 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN17 - tetraspanin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006616.2 | 520 | Silent Mutation | TCC,TCT | S,S 97 | NP_001006617.2 | |
NM_012171.2 | 520 | Silent Mutation | TCC,TCT | S,S 97 | NP_036303.1 | |
NM_130465.4 | 520 | Silent Mutation | TCC,TCT | S,S 97 | NP_569732.2 | |
XM_005265876.3 | 520 | Silent Mutation | TCC,TCT | S,S 31 | XP_005265933.1 |