Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAGCCAGAATAAAGATAAATGA[A/C]GAATTCAAAAATAATAAAAGTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYRM7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYRM7 - LYR motif containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293735.1 | 221 | Missense Mutation | GAA,GAC | E,D 38 | NP_001280664.1 | |
NM_181705.3 | 221 | Missense Mutation | GAA,GAC | E,D 38 | NP_859056.2 |