Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAGTTCATGAACTCCTCAGGGG[A/T]CACCTGCAGTGGAAAAGTTAGAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CAPSL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPSL - calcyphosine like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042625.1 | 606 | Missense Mutation | ACC,TCC | T,S 177 | NP_001036090.1 | |
NM_144647.3 | 606 | Missense Mutation | ACC,TCC | T,S 177 | NP_653248.3 | |
XM_006714444.3 | 606 | Missense Mutation | ACC,TCC | T,S 194 | XP_006714507.1 | |
XM_006714445.3 | 606 | UTR 3 | XP_006714508.1 |