Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGGCGGGAGAAAGAGCTCTCT[A/G]TACACTTTGTTCCCGGGAGCTGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605527 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAT2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAT2B - methionine adenosyltransferase 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013283.4 | 187 | Missense Mutation | ATA,GTA | I,V 10 | NP_037415.1 | |
NM_182796.2 | 187 | Intron | NP_877725.1 |