Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCTGACCTTCTCCACGTCGGCC[C/T]GGGCCGTCTGGTAATTGTCCACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604660 MIM: 603951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNIP1 - potassium voltage-gated channel interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034837.2 | 793 | Intron | NP_001030009.1 | |||
NM_001034838.2 | 793 | Intron | NP_001030010.1 | |||
NM_001278339.1 | 793 | Intron | NP_001265268.1 | |||
NM_001278340.1 | 793 | Intron | NP_001265269.1 | |||
NM_014592.3 | 793 | Intron | NP_055407.1 | |||
XM_017009407.1 | 793 | Intron | XP_016864896.1 | |||
XM_017009408.1 | 793 | Intron | XP_016864897.1 |
KCNMB1 - potassium calcium-activated channel subfamily M regulatory beta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004137.3 | 793 | Missense Mutation | CAG,CGG | Q,R 117 | NP_004128.1 |