Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGTCACCGCCAGTGCGGCCCCGG[A/G]GACCGCGAGCCTCGTCCCGGATTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114080 | ||||||||||||||||||||
Literature Links: |
CAMK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAMK4 - calcium/calmodulin dependent protein kinase IV | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323374.1 | 170 | Missense Mutation | GAG,GGG | E,G 24 | NP_001310303.1 | |
NM_001323375.1 | 170 | Missense Mutation | GAG,GGG | E,G 24 | NP_001310304.1 | |
NM_001323376.1 | 170 | UTR 5 | NP_001310305.1 | |||
NM_001323377.1 | 170 | UTR 5 | NP_001310306.1 | |||
NM_001744.5 | 170 | Missense Mutation | GAG,GGG | E,G 24 | NP_001735.1 |