Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTAACAGGACTCCAGCTTCATC[A/G]TCAGGTATGTGTTATGGGAAAAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605281 | ||||||||||||||||||||
Literature Links: |
DDX4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX4 - DEAD-box helicase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142549.1 | 245 | Silent Mutation | TCA,TCG | S,S 41 | NP_001136021.1 | |
NM_001166533.1 | 245 | Silent Mutation | TCA,TCG | S,S 41 | NP_001160005.1 | |
NM_001166534.1 | 245 | Intron | NP_001160006.1 | |||
NM_024415.2 | 245 | Silent Mutation | TCA,TCG | S,S 41 | NP_077726.1 | |
XM_011543495.2 | 245 | Silent Mutation | TCA,TCG | S,S 41 | XP_011541797.1 | |
XM_011543497.2 | 245 | Silent Mutation | TCA,TCG | S,S 41 | XP_011541799.1 |