Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCAGGCCATTTATGGCCTCCAT[C/G]CAGTATGGCGGACATCACGTTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600784 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GZMK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GZMK - granzyme K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002104.2 | 176 | Missense Mutation | ATC,ATG | I,M 44 | NP_002095.1 |
LOC102467081 - uncharacterized LOC102467081 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |