Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTGCATGTGCACCAGTATCACA[A/T]TATCGAGCTGTTGAAAGTGTGGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 107323 MIM: 604924 | ||||||||||||||||||||
Literature Links: |
ALDH7A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH7A1 - aldehyde dehydrogenase 7 family member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHAX - phosphorylated adaptor for RNA export | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032177.3 | Intron | NP_115553.2 |