Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACCTCGTTGGGGTCATCCTGGT[C/T]GGCATAGATTAGGAAGCCCGTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF2 - C1q and tumor necrosis factor related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031908.4 | 994 | Missense Mutation | AAC,GAC | N,D 323 | NP_114114.2 | |
XM_011534425.2 | 994 | Missense Mutation | AAC,GAC | N,D 362 | XP_011532727.1 | |
XM_011534426.2 | 994 | Missense Mutation | AAC,GAC | N,D 294 | XP_011532728.1 | |
XM_011534427.2 | 994 | UTR 3 | XP_011532729.1 | |||
XM_017009013.1 | 994 | Missense Mutation | AAC,GAC | N,D 255 | XP_016864502.1 |
CCNJL - cyclin J like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |