Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGTAGAAGTAGGGGTTTGGCTT[C/T]TGGCCCAGATCCACGCCCTTGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609450 MIM: 606681 MIM: 605733 MIM: 612415 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MXD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MXD3 - MAX dimerization protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NSD1 - nuclear receptor binding SET domain protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRELID1 - PRELI domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB24 - RAB24, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031677.3 | 943 | Silent Mutation | CAA,CAG | Q,Q 191 | NP_001026847.1 | |
NM_130781.3 | 943 | Silent Mutation | CAA,CAG | Q,Q 191 | NP_570137.2 |