Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCATGGTGCTCAAGGTGGCGGGG[A/C]CTGCATGTGCCGTGGTTGGGCTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC105379030 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105379030 - uncharacterized LOC105379030 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM171 - transmembrane protein 171 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161342.2 | 394 | Missense Mutation | ACT,CCT | T,P 64 | NP_001154814.1 | |
NM_173490.7 | 394 | Missense Mutation | ACT,CCT | T,P 64 | NP_775761.4 | |
XM_011543156.1 | 394 | Missense Mutation | ACT,CCT | T,P 64 | XP_011541458.1 |