Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCTGGGATCTTATCCATGTTTT[C/T]GGTTACTCTGCAAAACAGAGGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611197 | ||||||||||||||||||||
Literature Links: |
JAKMIP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JAKMIP2 - janus kinase and microtubule interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270934.1 | 2912 | UTR 3 | NP_001257863.1 | |||
NM_001270941.1 | 2912 | UTR 3 | NP_001257870.1 | |||
NM_001282282.1 | 2912 | UTR 3 | NP_001269211.1 | |||
NM_014790.4 | 2912 | Missense Mutation | CAA,CGA | Q,R 807 | NP_055605.2 | |
XM_017010097.1 | 2912 | UTR 3 | XP_016865586.1 | |||
XM_017010098.1 | 2912 | UTR 3 | XP_016865587.1 |
JAKMIP2-AS1 - JAKMIP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |