Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGATCCTAATCTTCAACAACCAC[A/G]GGAAGCCGCGGCTCTCCAAGTTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601507 MIM: 609608 | ||||||||||||||||||||
Literature Links: |
AP3S1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP3S1 - adaptor related protein complex 3 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002924.2 | 466 | UTR 5 | NP_001002924.2 | |||
NM_001284.3 | 466 | Missense Mutation | AGG,GGG | R,G 12 | NP_001275.1 | |
NM_001318090.1 | 466 | Missense Mutation | AGG,GGG | R,G 12 | NP_001305019.1 | |
NM_001318091.1 | 466 | Missense Mutation | AGG,GGG | R,G 12 | NP_001305020.1 | |
NM_001318093.1 | 466 | Missense Mutation | AGG,GGG | R,G 12 | NP_001305022.1 | |
NM_001318094.1 | 466 | Missense Mutation | AGG,GGG | R,G 12 | NP_001305023.1 | |
XM_011543136.2 | 466 | Missense Mutation | AGG,GGG | R,G 112 | XP_011541438.2 | |
XM_017009024.1 | 466 | Missense Mutation | AGG,GGG | R,G 112 | XP_016864513.1 | |
XM_017009025.1 | 466 | Missense Mutation | AGG,GGG | R,G 112 | XP_016864514.1 |
ATG12 - autophagy related 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277783.1 | 466 | Intron | NP_001264712.1 | |||
NM_004707.3 | 466 | Intron | NP_004698.3 |