Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGATTCGGCAGATGCACCCAAC[C/T]CCTGGTCGAGGGAGCCGCGTGACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610530 | ||||||||||||||||||||
Literature Links: |
MIR4638 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4638 - microRNA 4638 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM41 - tripartite motif containing 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033549.4 | 1377 | Silent Mutation | ACC,ACT | T,T 214 | NP_291027.3 | |
NM_201627.2 | 1377 | Silent Mutation | ACC,ACT | T,T 214 | NP_963921.1 | |
XM_006714929.2 | 1377 | Silent Mutation | ACC,ACT | T,T 214 | XP_006714992.1 | |
XM_006714930.2 | 1377 | Silent Mutation | ACC,ACT | T,T 214 | XP_006714993.1 | |
XM_006714931.3 | 1377 | Silent Mutation | ACC,ACT | T,T 214 | XP_006714994.1 | |
XM_006714932.3 | 1377 | Silent Mutation | ACC,ACT | T,T 214 | XP_006714995.1 |