Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGTACCTTTCACAATTTAAAAC[C/T]TGGCTGGGATTCTCAACATATCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606694 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP155 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUP155 - nucleoporin 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278312.1 | 4169 | Silent Mutation | CAA,CAG | Q,Q 1276 | NP_001265241.1 | |
NM_004298.3 | 4169 | Silent Mutation | CAA,CAG | Q,Q 1281 | NP_004289.1 | |
NM_153485.2 | 4169 | Silent Mutation | CAA,CAG | Q,Q 1340 | NP_705618.1 | |
XM_011514165.2 | 4169 | Intron | XP_011512467.1 |