Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTCCCTACGTTGCTTGGTAAA[C/T]ATATTTGCTTCTCCTTTCTGCAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIOK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RIOK2 - RIO kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159749.1 | 1694 | Intron | NP_001153221.1 | |||
NM_018343.2 | 1694 | Missense Mutation | ATA,ATG | I,M 528 | NP_060813.2 | |
XM_017009628.1 | 1694 | Missense Mutation | ATA,ATG | I,M 341 | XP_016865117.1 |