Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTTTTTTCCAGGTTTATGATG[A/T]TAAAGACTACAGGAGAATTCGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611842 MIM: 603848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL36 - mitochondrial ribosomal protein L36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032479.3 | 179 | Intron | NP_115868.1 | |||
XM_011514079.2 | 179 | Intron | XP_011512381.1 | |||
XM_011514080.2 | 179 | Intron | XP_011512382.1 | |||
XM_017009750.1 | 179 | Intron | XP_016865239.1 | |||
XM_017009751.1 | 179 | Intron | XP_016865240.1 | |||
XM_017009752.1 | 179 | Intron | XP_016865241.1 |
NDUFS6 - NADH:ubiquinone oxidoreductase subunit S6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004553.4 | 179 | Missense Mutation | GAT,GTT | D,V 48 | NP_004544.1 |