Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGATTTCGGGCCCCTCGATATGCG[A/G]TAACAACAAACCATGGTAGTCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606501 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTMR12 - myotubularin related protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040446.2 | 1725 | Missense Mutation | CCG,TCG | P,S 629 | NP_001035536.1 | |
NM_001294343.1 | 1725 | Missense Mutation | CCG,TCG | P,S 575 | NP_001281272.1 | |
NM_001294344.1 | 1725 | Missense Mutation | CCG,TCG | P,S 519 | NP_001281273.1 | |
XM_005248313.3 | 1725 | Missense Mutation | CCG,TCG | P,S 649 | XP_005248370.1 | |
XM_011514057.1 | 1725 | Missense Mutation | CCG,TCG | P,S 681 | XP_011512359.1 | |
XM_011514058.2 | 1725 | Missense Mutation | CCG,TCG | P,S 661 | XP_011512360.1 | |
XM_011514059.1 | 1725 | Missense Mutation | CCG,TCG | P,S 625 | XP_011512361.1 | |
XM_011514060.1 | 1725 | Missense Mutation | CCG,TCG | P,S 551 | XP_011512362.1 | |
XM_017009589.1 | 1725 | Missense Mutation | CCG,TCG | P,S 573 | XP_016865078.1 | |
XM_017009590.1 | 1725 | Missense Mutation | CCG,TCG | P,S 478 | XP_016865079.1 | |
XM_017009591.1 | 1725 | Missense Mutation | CCG,TCG | P,S 478 | XP_016865080.1 | |
XM_017009592.1 | 1725 | Missense Mutation | CCG,TCG | P,S 446 | XP_016865081.1 | |
XM_017009593.1 | 1725 | Missense Mutation | CCG,TCG | P,S 426 | XP_016865082.1 | |
XM_017009594.1 | 1725 | Missense Mutation | CCG,TCG | P,S 426 | XP_016865083.1 | |
XM_017009595.1 | 1725 | Missense Mutation | CCG,TCG | P,S 426 | XP_016865084.1 | |
XM_017009596.1 | 1725 | Missense Mutation | CCG,TCG | P,S 370 | XP_016865085.1 |