Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGTGGGATGGTACAGGCTCACTC[A/G]CAGGACGGTGAGGGGCTTCTCCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609710 MIM: 612663 | ||||||||||||||||||||
Literature Links: |
DCANP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCANP1 - dendritic cell associated nuclear protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIFAB - TIFA inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099221.1 | 226 | Nonsense Mutation | CGA,TGA | R,* 9 | NP_001092691.1 |