Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCTCAGTCTACTCGACTCCTGT[A/G]TTTTCACAGAAAATGACGGTGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137164 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRG2 - gamma-aminobutyric acid type A receptor gamma2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000816.3 | 412 | Silent Mutation | GTA,GTG | V,V 18 | NP_000807.2 | |
NM_198903.2 | 412 | Silent Mutation | GTA,GTG | V,V 18 | NP_944493.2 | |
NM_198904.2 | 412 | Silent Mutation | GTA,GTG | V,V 18 | NP_944494.1 |