Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAACTGGTAGGCCTGCTAGAAG[A/T]TGCTGATTGCTGGTATCAAGGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602226 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD180 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD180 - CD180 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005582.2 | 1546 | Missense Mutation | CAA,CAT | Q,H 463 | NP_005573.2 | |
XM_005248504.4 | 1546 | Missense Mutation | CAA,CAT | Q,H 450 | XP_005248561.1 |