Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTGAGCCTGTGCAGCGAGCAG[A/G]ACATCGAGGGCTGGCTCGAGGGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SNX18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX18 - sorting nexin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102575.1 | 290 | Missense Mutation | AAC,GAC | N,D 34 | NP_001096045.1 | |
NM_001145427.1 | 290 | Missense Mutation | AAC,GAC | N,D 34 | NP_001138899.1 | |
NM_052870.2 | 290 | Missense Mutation | AAC,GAC | N,D 34 | NP_443102.2 | |
XM_017008997.1 | 290 | Missense Mutation | AAC,GAC | N,D 34 | XP_016864486.1 |