Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGTTGCTGTTCTCTCTGGATC[A/C]ACTGTACAAGGTCTAGATGCCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611989 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPS27 - mitochondrial ribosomal protein S27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286748.1 | 1170 | Missense Mutation | TGG,TTG | W,L 403 | NP_001273677.1 | |
NM_001286751.1 | 1170 | Missense Mutation | TGG,TTG | W,L 333 | NP_001273680.1 | |
NM_015084.2 | 1170 | Missense Mutation | TGG,TTG | W,L 389 | NP_055899.2 |