Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCACGCAGCCACCTGGCCACCA[C/T]ACCCAGCAGCTTCCCACAAAGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616383 | ||||||||||||||||||||
Literature Links: |
UGT3A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UGT3A1 - UDP glycosyltransferase family 3 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171873.1 | 1447 | Intron | NP_001165344.1 | |||
NM_152404.3 | 1447 | Missense Mutation | ATG,GTG | M,V 509 | NP_689617.3 | |
XM_005248243.4 | 1447 | Missense Mutation | ATG,GTG | M,V 455 | XP_005248300.1 | |
XM_011513957.2 | 1447 | Missense Mutation | ATG,GTG | M,V 478 | XP_011512259.1 | |
XM_011513958.2 | 1447 | Missense Mutation | ATG,GTG | M,V 475 | XP_011512260.1 | |
XM_011513959.2 | 1447 | Missense Mutation | ATG,GTG | M,V 455 | XP_011512261.1 | |
XM_011513961.2 | 1447 | Missense Mutation | ATG,GTG | M,V 396 | XP_011512263.1 |