Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGGCCGCGGGCGGCGGCCCGCCC[G/T]CAGGGAAGTCCGGGGCAGCGTCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602406 | ||||||||||||||||||||
Literature Links: |
HAND1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HAND1 - heart and neural crest derivatives expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004821.2 | 374 | Missense Mutation | GAG,GCG | E,A 58 | NP_004812.1 | |
XM_005268531.1 | 374 | Missense Mutation | GAG,GCG | E,A 58 | XP_005268588.1 |