Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGAAGCGGGAGCTGGCTGGGGA[C/G]CAGCCGTACCGCCGCGCTCTGCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 122559 MIM: 610103 | ||||||||||||||||||||
Literature Links: |
CRHBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRHBP - corticotropin releasing hormone binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001882.3 | 433 | Missense Mutation | GAC,GAG | D,E 51 | NP_001873.2 |
S100Z - S100 calcium binding protein Z | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130772.3 | 433 | Intron | NP_570128.2 | |||
XM_011543239.2 | 433 | Intron | XP_011541541.1 | |||
XM_011543240.2 | 433 | Intron | XP_011541542.1 | |||
XM_011543241.2 | 433 | Intron | XP_011541543.1 | |||
XM_011543244.2 | 433 | Intron | XP_011541546.1 | |||
XM_011543245.2 | 433 | Intron | XP_011541547.1 | |||
XM_017009171.1 | 433 | Intron | XP_016864660.1 | |||
XM_017009172.1 | 433 | Intron | XP_016864661.1 | |||
XM_017009173.1 | 433 | Intron | XP_016864662.1 |