Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGATGATGTCCCCTGTGCAGAA[C/T]GGTGTACCTAGAAAGGAAGGAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608533 MIM: 602164 | ||||||||||||||||||||
Literature Links: |
FBXO38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO38 - F-box protein 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HTR4 - 5-hydroxytryptamine receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000870.6 | 1207 | Intron | NP_000861.1 | |||
NM_001040169.2 | 1207 | Missense Mutation | ATT,GTT | I,V 362 | NP_001035259.1 | |
NM_001040172.2 | 1207 | Intron | NP_001035262.2 | |||
NM_001040173.2 | 1207 | Intron | NP_001035263.1 | |||
NM_001286410.1 | 1207 | Intron | NP_001273339.1 | |||
NM_199453.3 | 1207 | UTR 3 | NP_955525.1 |