Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGCAAGATGAAGGTGGAGCCA[A/G]ATGACACAATCTGGTCTCACTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138321 MIM: 607714 | ||||||||||||||||||||
Literature Links: |
GPX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPX3 - glutathione peroxidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNIP1 - TNFAIP3 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252385.1 | 2034 | UTR 3 | NP_001239314.1 | |||
NM_001252386.1 | 2034 | UTR 3 | NP_001239315.1 | |||
NM_001252390.1 | 2034 | UTR 3 | NP_001239319.1 | |||
NM_001252391.1 | 2034 | UTR 3 | NP_001239320.1 | |||
NM_001252392.1 | 2034 | Intron | NP_001239321.1 | |||
NM_001252393.1 | 2034 | Intron | NP_001239322.1 | |||
NM_001258454.1 | 2034 | UTR 3 | NP_001245383.1 | |||
NM_001258455.1 | 2034 | UTR 3 | NP_001245384.1 | |||
NM_001258456.1 | 2034 | Silent Mutation | CTG,TTG | L,L 547 | NP_001245385.1 | |
NM_006058.4 | 2034 | UTR 3 | NP_006049.3 | |||
XM_005268355.1 | 2034 | Intron | XP_005268412.1 | |||
XM_006714751.1 | 2034 | UTR 3 | XP_006714814.1 | |||
XM_006714752.2 | 2034 | Intron | XP_006714815.1 | |||
XM_017008945.1 | 2034 | Intron | XP_016864434.1 | |||
XM_017008946.1 | 2034 | Intron | XP_016864435.1 | |||
XM_017008947.1 | 2034 | Intron | XP_016864436.1 | |||
XM_017008948.1 | 2034 | Intron | XP_016864437.1 | |||
XM_017008949.1 | 2034 | Intron | XP_016864438.1 | |||
XM_017008950.1 | 2034 | Intron | XP_016864439.1 | |||
XM_017008951.1 | 2034 | Intron | XP_016864440.1 |