Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCGGGTGATTATTCATGATAAC[C/T]GCATCACACAACGAATCTATGAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606698 | ||||||||||||||||||||
Literature Links: |
C5orf52 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C5orf52 - chromosome 5 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145132.1 | 354 | Missense Mutation | CGC,TGC | R,C 98 | NP_001138604.1 | |
XM_011534416.1 | 354 | Missense Mutation | CGC,TGC | R,C 98 | XP_011532718.1 | |
XM_017008909.1 | 354 | Intron | XP_016864398.1 |
SOX30 - SRY-box 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |