Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAGGGCTACCATGAGATGGGAC[A/G]GGCAGCAGCTGTCCTCCTCAGTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610207 | ||||||||||||||||||||
Literature Links: |
SLC4A9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC4A9 - solute carrier family 4 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258426.1 | 814 | Missense Mutation | CAG,CGG | Q,R 260 | NP_001245355.1 | |
NM_001258427.1 | 814 | Missense Mutation | CAG,CGG | Q,R 260 | NP_001245356.1 | |
NM_001258428.1 | 814 | Missense Mutation | CAG,CGG | Q,R 284 | NP_001245357.1 | |
NM_031467.2 | 814 | Missense Mutation | CAG,CGG | Q,R 260 | NP_113655.2 | |
XM_005268521.3 | 814 | Missense Mutation | CAG,CGG | Q,R 284 | XP_005268578.2 | |
XM_011537694.2 | 814 | Missense Mutation | CAG,CGG | Q,R 308 | XP_011535996.1 | |
XM_017009933.1 | 814 | Missense Mutation | CAG,CGG | Q,R 308 | XP_016865422.1 | |
XM_017009934.1 | 814 | Missense Mutation | CAG,CGG | Q,R 308 | XP_016865423.1 | |
XM_017009935.1 | 814 | Missense Mutation | CAG,CGG | Q,R 308 | XP_016865424.1 | |
XM_017009936.1 | 814 | Missense Mutation | CAG,CGG | Q,R 284 | XP_016865425.1 | |
XM_017009937.1 | 814 | Missense Mutation | CAG,CGG | Q,R 270 | XP_016865426.1 | |
XM_017009938.1 | 814 | Missense Mutation | CAG,CGG | Q,R 308 | XP_016865427.1 | |
XM_017009939.1 | 814 | Missense Mutation | CAG,CGG | Q,R 284 | XP_016865428.1 | |
XM_017009940.1 | 814 | Missense Mutation | CAG,CGG | Q,R 308 | XP_016865429.1 | |
XM_017009941.1 | 814 | Missense Mutation | CAG,CGG | Q,R 308 | XP_016865430.1 | |
XM_017009942.1 | 814 | Missense Mutation | CAG,CGG | Q,R 308 | XP_016865431.1 | |
XM_017009943.1 | 814 | Missense Mutation | CAG,CGG | Q,R 246 | XP_016865432.1 |