Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGGCCTATCTGTCAAAGCAGC[C/T]GGTGGGGAACATGTGGCTCCCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNJL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNJL - cyclin J like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308173.1 | 1212 | Missense Mutation | AGC,GGC | S,G 383 | NP_001295102.1 | |
NM_024565.6 | 1212 | Missense Mutation | AGC,GGC | S,G 431 | NP_078841.3 | |
XM_006714917.3 | 1212 | Missense Mutation | AGC,GGC | S,G 428 | XP_006714980.1 | |
XM_011534646.2 | 1212 | Missense Mutation | AGC,GGC | S,G 361 | XP_011532948.1 | |
XM_011534647.2 | 1212 | Missense Mutation | AGC,GGC | S,G 325 | XP_011532949.1 | |
XM_017009846.1 | 1212 | Missense Mutation | AGC,GGC | S,G 533 | XP_016865335.1 | |
XM_017009847.1 | 1212 | Missense Mutation | AGC,GGC | S,G 384 | XP_016865336.1 | |
XM_017009848.1 | 1212 | Missense Mutation | AGC,GGC | S,G 313 | XP_016865337.1 | |
XM_017009849.1 | 1212 | Missense Mutation | AGC,GGC | S,G 117 | XP_016865338.1 |