Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACCTGCTGAGAGGCAACAAGG[A/G]CAGCAGAGGTCAAGCCTAATGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF608 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF608 - zinc finger protein 608 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020747.2 | 3837 | Missense Mutation | GCC,GTC | A,V 1489 | NP_065798.2 | |
XM_005272037.2 | 3837 | Missense Mutation | GCC,GTC | A,V 1489 | XP_005272094.1 | |
XM_005272038.1 | 3837 | Missense Mutation | GCC,GTC | A,V 1489 | XP_005272095.1 | |
XM_005272039.2 | 3837 | Missense Mutation | GCC,GTC | A,V 1489 | XP_005272096.1 | |
XM_005272040.3 | 3837 | Missense Mutation | GCC,GTC | A,V 1201 | XP_005272097.1 | |
XM_011543520.2 | 3837 | UTR 3 | XP_011541822.1 | |||
XM_011543522.2 | 3837 | Missense Mutation | GCC,GTC | A,V 1192 | XP_011541824.1 | |
XM_011543523.2 | 3837 | Missense Mutation | GCC,GTC | A,V 1104 | XP_011541825.1 | |
XM_017009674.1 | 3837 | Missense Mutation | GCC,GTC | A,V 1489 | XP_016865163.1 |