Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAACAAAAGACAGGTGAGACTT[C/T]GAGAGCCCTGTCATCTTCTAAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUB1 - SUB1 homolog, transcriptional regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006713.3 | 518 | Intron | NP_006704.3 | |||
XM_011513944.2 | 518 | Missense Mutation | TCG,TTG | S,L 46 | XP_011512246.1 | |
XM_017008986.1 | 518 | Intron | XP_016864475.1 | |||
XM_017008987.1 | 518 | Intron | XP_016864476.1 |