Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGCTGGCCCCTGGAGGAGTTCC[C/G]GCTGGGAGCGCTGCTGGGTCCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC14B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC14B - leucine rich repeat containing 14B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080478.1 | 216 | Missense Mutation | CCG,CGG | P,R 63 | NP_001073947.1 |
PLEKHG4B - pleckstrin homology and RhoGEF domain containing G4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |