Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGAACAAACCCCGCTGCGTCTG[C/T]GCCCCGGATTGTTCCAACATCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136470 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FST - follistatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006350.3 | 522 | Silent Mutation | TGC,TGT | C,C 118 | NP_006341.1 | |
NM_013409.2 | 522 | Silent Mutation | TGC,TGT | C,C 118 | NP_037541.1 | |
XM_005248400.4 | 522 | Silent Mutation | TGC,TGT | C,C 118 | XP_005248457.1 | |
XM_005248401.4 | 522 | Silent Mutation | TGC,TGT | C,C 118 | XP_005248458.1 | |
XM_005248402.4 | 522 | Silent Mutation | TGC,TGT | C,C 118 | XP_005248459.1 | |
XM_005248403.4 | 522 | Silent Mutation | TGC,TGT | C,C 118 | XP_005248460.1 | |
XM_011543099.2 | 522 | Silent Mutation | TGC,TGT | C,C 113 | XP_011541401.1 | |
XM_017008954.1 | 522 | Silent Mutation | TGC,TGT | C,C 118 | XP_016864443.1 | |
XM_017008955.1 | 522 | Silent Mutation | TGC,TGT | C,C 40 | XP_016864444.1 |