Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGTTGAAAAATGCCAACTTCAC[C/T]GGCCCCAACCAGACCTCGAGCAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104220 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADRA1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADRA1B - adrenoceptor alpha 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000679.3 | 251 | Silent Mutation | ACC,ACT | T,T 26 | NP_000670.1 | |
XM_005265818.3 | 251 | Silent Mutation | ACC,ACT | T,T 26 | XP_005265875.1 | |
XM_005265819.2 | 251 | Silent Mutation | ACC,ACT | T,T 26 | XP_005265876.1 | |
XM_006714821.3 | 251 | Silent Mutation | ACC,ACT | T,T 26 | XP_006714884.1 | |
XM_011534435.1 | 251 | Silent Mutation | ACC,ACT | T,T 26 | XP_011532737.1 | |
XM_011534437.2 | 251 | Silent Mutation | ACC,ACT | T,T 26 | XP_011532739.1 | |
XM_011534438.2 | 251 | Silent Mutation | ACC,ACT | T,T 26 | XP_011532740.1 |